Product Details

SNP ID
rs113707187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42387646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGATGGCTTAGCGATTGGAAG[A/G]CAGGGTTTTTCTTTTTTTTTTTTTC
Phenotype
MIM: 611764 MIM: 604267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNFN PubMed Links

Gene Details

Gene
CNFN
Gene Name
cornifelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032488.3 Intron NP_115877.2
XM_005259332.2 Intron XP_005259389.1
XM_011527396.1 Intron XP_011525698.1
XM_011527397.1 Intron XP_011525699.1
Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC101930071
Gene Name
uncharacterized LOC101930071
There are no transcripts associated with this gene.

Gene
MEGF8
Gene Name
multiple EGF like domains 8
There are no transcripts associated with this gene.

View Full Product Details