Product Details

SNP ID
rs115712534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46079488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGCGGATTGGAGCCTGTTTGCA[C/T]CCCTATAGCCCGAATGAGACCCAAG
Phenotype
MIM: 610545
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFL2 PubMed Links

Gene Details

Gene
IGFL2
Gene Name
IGF like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002915.2 Intron NP_001002915.2
NM_001135113.1 Intron NP_001128585.1
XM_011526501.1 Intron XP_011524803.1
XM_017026335.1 Intron XP_016881824.1
Gene
LOC400706
Gene Name
uncharacterized LOC400706
There are no transcripts associated with this gene.

View Full Product Details