Product Details

SNP ID
rs112364896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46648160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTAGGCTTCTAAACTAAAACGGG[G/A]AAATTCAAACCGAATTACCCCTTTT
Phenotype
MIM: 611112
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DACT3 PubMed Links

Gene Details

Gene
DACT3
Gene Name
dishevelled binding antagonist of beta catenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301046.1 3062 UTR 3 NP_001287975.1
NM_145056.2 3062 UTR 3 NP_659493.2
XM_011526499.2 3062 UTR 3 XP_011524801.1
XM_011526500.2 3062 UTR 3 XP_011524802.1
XM_017026334.1 3062 UTR 3 XP_016881823.1
Gene
GNG8
Gene Name
G protein subunit gamma 8
There are no transcripts associated with this gene.

View Full Product Details