Product Details

SNP ID
rs112829612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49890149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAACGCCTCTTCCCGGCTCAAGC[C/G]GGCGAACGCTGCCGCCGCGTCCGAC
Phenotype
MIM: 609742 MIM: 616659
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL4I1 PubMed Links

Gene Details

Gene
IL4I1
Gene Name
interleukin 4 induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258017.1 1854 Missense Mutation CGC,GGC R431G NP_001244946.1
NM_001258018.1 1854 Missense Mutation CGC,GGC R431G NP_001244947.1
NM_152899.1 1854 Missense Mutation CGC,GGC R409G NP_690863.1
NM_172374.2 1854 Missense Mutation CGC,GGC R431G NP_758962.1
Gene
MIR4750
Gene Name
microRNA 4750
There are no transcripts associated with this gene.

Gene
TBC1D17
Gene Name
TBC1 domain family member 17
There are no transcripts associated with this gene.

View Full Product Details