Product Details

SNP ID
rs117644723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11723076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCGGTGGGGGGCCTTTCATGT[C/G]TCTGGAAGGAGTGCTGATGACTGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 521 Missense Mutation ACA,AGA T153R NP_001073962.1
NM_001297610.1 521 UTR 5 NP_001284539.1
NM_017507.1 521 Missense Mutation ACA,AGA T109R NP_059977.1
XM_017026941.1 521 UTR 5 XP_016882430.1
XM_017026942.1 521 UTR 5 XP_016882431.1

View Full Product Details