Product Details

SNP ID
rs113467048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2463134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATAGGTGGGGGAGGACCCCCAC[A/G]GCCAGGTGGGTACTGGGTTGGAGCC
Phenotype
MIM: 182282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORD119 PubMed Links

Gene Details

Gene
SNORD119
Gene Name
small nucleolar RNA, C/D box 119
There are no transcripts associated with this gene.

Gene
SNRPB
Gene Name
small nuclear ribonucleoprotein polypeptides B and B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003091.3 677 Missense Mutation CGT,TGT R172C NP_003082.1
NM_198216.1 677 Missense Mutation CGT,TGT R172C NP_937859.1

View Full Product Details