Product Details
- SNP ID
-
rs113156985
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:157553 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGGAAAGCCTGGCCATGGGGCT[C/T]TTCATGATCATTGCAATTCTGCTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB127
PubMed Links
Gene Details
- Gene
- DEFB127
- Gene Name
- defensin beta 127
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139074.3 |
84 |
Silent Mutation |
CTC,CTT |
L3L |
NP_620713.1 |
View Full Product Details