Product Details

SNP ID
rs112897665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30812904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCTGGGACTCGAGGTAAGTTG[G/C]AAAGCAGTGTCTCTGAGGTTGATGA
Phenotype
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP8-1 PubMed Links

Gene Details

Gene
KRTAP8-1
Gene Name
keratin associated protein 8-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175857.3 349 UTR 3 NP_787053.1

View Full Product Details