Product Details

SNP ID
rs117254218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36328299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGTATTTCCTGATAACTATGA[G/A]TGGAACATCTTTTGTTATTTATAAT
Phenotype
MIM: 610078
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MORC3 PubMed Links

Gene Details

Gene
MORC3
Gene Name
MORC family CW-type zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320445.1 Intron NP_001307374.1
NM_001320446.1 Intron NP_001307375.1
NM_015358.2 Intron NP_056173.1
XM_011529518.2 Intron XP_011527820.1
XM_017028311.1 Intron XP_016883800.1
XM_017028312.1 Intron XP_016883801.1

View Full Product Details