Product Details

SNP ID
rs113814620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74414814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGATAGGCCTAAATTTAAAGTGG[G/T]GAACAGGAGTGCGGGGCCCGAGTCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145054.1 1433 Missense Mutation ACC,CCC T428P NP_001138526.1
NM_001316764.1 1433 Missense Mutation ACC,CCC T455P NP_001303693.1
NM_001316765.1 1433 Missense Mutation ACC,CCC T406P NP_001303694.1
NM_001316766.1 1433 Missense Mutation ACC,CCC T360P NP_001303695.1
Gene
WDR54
Gene Name
WD repeat domain 54
There are no transcripts associated with this gene.

View Full Product Details