Product Details

SNP ID
rs116683909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14404403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGATCCAAAGGGGGCCTAGCAC[A/G]GCTAAGAACCCCCAACAGGGTGGGA
Phenotype
MIM: 186854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A6 PubMed Links

Gene Details

Gene
SLC6A6
Gene Name
solute carrier family 6 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134367.3 Intron NP_001127839.2
NM_001134368.3 Intron NP_001127840.1
NM_003043.5 Intron NP_003034.2
XM_006713307.2 Intron XP_006713370.1
XM_011534030.2 Intron XP_011532332.1
XM_011534031.2 Intron XP_011532333.1

View Full Product Details