Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001144026.2 | Intron | NP_001137498.1 | |||
NM_001144027.2 | Intron | NP_001137499.1 | |||
NM_001144028.2 | Intron | NP_001137500.1 | |||
NM_014434.3 | Intron | NP_055249.1 | |||
XM_011518545.1 | Intron | XP_011516847.1 | |||
XM_011518547.1 | Intron | XP_011516849.1 | |||
XM_011518548.2 | Intron | XP_011516850.1 | |||
XM_017014626.1 | Intron | XP_016870115.1 | |||
XM_017014627.1 | Intron | XP_016870116.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_053045.1 | Intron | NP_444273.1 |