Product Details

SNP ID
rs116374352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127708242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCAACCTTCCCAAATCTCCAA[C/T]AGGTGAGCATGAAGTTTATTACACT
Phenotype
MIM: 613607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THEMIS PubMed Links

Gene Details

Gene
THEMIS
Gene Name
thymocyte selection associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010923.2 3935 UTR 3 NP_001010923.1
NM_001164685.1 3935 UTR 3 NP_001158157.1
NM_001164687.1 3935 UTR 3 NP_001158159.1
NM_001318531.1 3935 UTR 3 NP_001305460.1
XM_011535813.2 3935 UTR 3 XP_011534115.1
XM_011535814.2 3935 UTR 3 XP_011534116.1
XM_011535816.1 3935 UTR 3 XP_011534118.1
XM_017010848.1 3935 Intron XP_016866337.1
XM_017010849.1 3935 UTR 3 XP_016866338.1

View Full Product Details