Product Details

SNP ID
rs112515167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99435772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTGCACTAAGTGCTCTTGATT[C/G]TGGAACCACCTGTAAGTAAGTGTCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927365 PubMed Links

Gene Details

Gene
LOC101927365
Gene Name
uncharacterized LOC101927365
There are no transcripts associated with this gene.

Gene
TSTD3
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195131.1 1805 Intron NP_001182060.1
XM_017010141.1 1805 Intron XP_016865630.1
Gene
USP45
Gene Name
ubiquitin specific peptidase 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080481.1 1805 Missense Mutation CAA,GAA Q797E NP_001073950.1
XM_005267169.3 1805 Missense Mutation CAA,GAA Q797E XP_005267226.1
XM_005267170.4 1805 Missense Mutation CAA,GAA Q797E XP_005267227.1
XM_005267172.3 1805 UTR 3 XP_005267229.1
XM_005267175.3 1805 Intron XP_005267232.1
XM_011536200.2 1805 Missense Mutation CAA,GAA Q797E XP_011534502.1
XM_017011381.1 1805 Missense Mutation CAA,GAA Q451E XP_016866870.1
XM_017011382.1 1805 Missense Mutation CAA,GAA Q437E XP_016866871.1
XM_017011383.1 1805 Missense Mutation CAA,GAA Q437E XP_016866872.1
XM_017011384.1 1805 Missense Mutation CAA,GAA Q437E XP_016866873.1
XM_017011385.1 1805 Intron XP_016866874.1
XM_017011386.1 1805 Intron XP_016866875.1

View Full Product Details