Product Details

SNP ID
rs113745698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:63598690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGCTAACTGGCTTGTTTTTACA[T/C]AAAGCTTTAAACATGTCTACCATGG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
WDR89 PubMed Links

Gene Details

Gene
WDR89
Gene Name
WD repeat domain 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008726.2 2015 UTR 3 NP_001008726.1
NM_001258272.1 2015 UTR 3 NP_001245201.1
NM_080666.3 2015 UTR 3 NP_542397.1
XM_011536383.2 2015 UTR 3 XP_011534685.1
XM_011536385.2 2015 UTR 3 XP_011534687.1

View Full Product Details