Product Details

SNP ID
rs112896997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150222404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTAGCCTACAGCCCAAGACAG[A/G]TCAGGCGGACAGCAGCTGAGGCTGG
Phenotype
MIM: 114078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMK2A PubMed Links

Gene Details

Gene
CAMK2A
Gene Name
calcium/calmodulin dependent protein kinase II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015981.3 1958 UTR 3 NP_057065.2
NM_171825.2 1958 UTR 3 NP_741960.1
XM_017009898.1 1958 Intron XP_016865387.1

View Full Product Details