Product Details
- SNP ID
-
rs113836536
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:142001275 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAACAGAGGAGCCACAGTGAGGC[C/T]TTCACAAAAACACAGGGCTCTGGGG
- Phenotype
-
MIM: 601798
MIM: 605675
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNPDA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs55892272] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNPDA1
- Gene Name
- glucosamine-6-phosphate deaminase 1
- Gene
- RNF14
- Gene Name
- ring finger protein 14
There are no transcripts associated with this gene.
View Full Product Details