Product Details

SNP ID
rs113208516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125495513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCGTCTGTAGTAAAACAATCG[C/T]TCTCCCGGCGTCTGCGGCCGCTGCC
Phenotype
MIM: 604825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FEZ1 PubMed Links

Gene Details

Gene
FEZ1
Gene Name
fasciculation and elongation protein zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005103.4 300 Intron NP_005094.1
NM_022549.3 300 Intron NP_072043.1
XM_005271734.2 300 UTR 5 XP_005271791.1
XM_005271735.2 300 Intron XP_005271792.1
Gene
LOC403312
Gene Name
putative uncharacterized protein MGC39545
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301851.1 300 Missense Mutation GCT,GTT A62V NP_001288780.1

View Full Product Details