Product Details

SNP ID
rs114976399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:128084635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTGTGCCCAGGATCGATCACTA[C/T]CGGCACACAGCCGCGCAGCTGGGCG
Phenotype
MIM: 600840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01184 PubMed Links

Gene Details

Gene
LINC01184
Gene Name
long intergenic non-protein coding RNA 1184
There are no transcripts associated with this gene.

Gene
SLC12A2
Gene Name
solute carrier family 12 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001046.2 845 Silent Mutation TAC,TAT Y227Y NP_001037.1
NM_001256461.1 845 Silent Mutation TAC,TAT Y227Y NP_001243390.1
XM_011543588.2 845 Silent Mutation TAC,TAT Y227Y XP_011541890.1
XM_017009771.1 845 UTR 5 XP_016865260.1

View Full Product Details