Product Details

SNP ID
rs112483111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:96936955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGAACGGGGAGCATAAATCCC[A/C]CCCTAGTTAGAACCATGGCTTTTGT
Phenotype
MIM: 606832 MIM: 151300
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ERAP1 PubMed Links

Gene Details

Gene
ERAP1
Gene Name
endoplasmic reticulum aminopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040458.1 Intron NP_001035548.1
NM_001198541.1 Intron NP_001185470.1
NM_016442.3 Intron NP_057526.3
XM_005272015.4 Intron XP_005272072.1
XM_005272016.4 Intron XP_005272073.1
XM_011543480.2 Intron XP_011541782.1
XM_011543481.2 Intron XP_011541783.1
XM_011543484.2 Intron XP_011541786.1
XM_011543485.2 Intron XP_011541787.1
XM_011543486.2 Intron XP_011541788.1
XM_017009580.1 Intron XP_016865069.1
XM_017009581.1 Intron XP_016865070.1
XM_017009582.1 Intron XP_016865071.1
XM_017009583.1 Intron XP_016865072.1
Gene
LNPEP
Gene Name
leucyl and cystinyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005575.2 Intron NP_005566.2
NM_175920.3 Intron NP_787116.2
Gene
LOC101929747
Gene Name
uncharacterized LOC101929747
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005272150.4 Intron XP_005272207.1

View Full Product Details