Product Details
- SNP ID
-
rs201847938
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:79559428 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACACAAACGTCCTTCACTTCGCAC[G/A]CAGCACCAGAGGCTGCCATCAAGAT
- Phenotype
-
MIM: 178642
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFTPA2
PubMed Links
Gene Details
- Gene
- SFTPA2
- Gene Name
- surfactant protein A2
There are no transcripts associated with this gene.
View Full Product Details