Product Details

SNP ID
rs113047876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133352082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCTGTTCCCTGTCCAGAAGCC[A/G]AAACTCTTGACCACCGAGGAGAAGC
Phenotype
MIM: 609776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUOM PubMed Links

Gene Details

Gene
FUOM
Gene Name
fucose mutarotase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098483.2 282 Intron NP_001091953.1
NM_001301827.1 282 Intron NP_001288756.1
NM_001301828.1 282 Intron NP_001288757.1
NM_198472.2 282 Intron NP_940874.2
XM_011539670.2 282 Intron XP_011537972.1
XM_011539671.2 282 Intron XP_011537973.1
XM_011539672.2 282 Intron XP_011537974.1
XM_011539673.2 282 Intron XP_011537975.1
XM_011539674.2 282 Intron XP_011537976.1
XM_011539675.2 282 Intron XP_011537977.1
XM_017016094.1 282 Intron XP_016871583.1
Gene
PRAP1
Gene Name
proline rich acidic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145201.1 282 Silent Mutation CCA,CCG P68P NP_001138673.1
NM_145202.4 282 Silent Mutation CCA,CCG P68P NP_660203.3

View Full Product Details