Product Details

SNP ID
rs116351032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:35256157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTTTATCTATGCTGCTAATCTC[T/C]GATTTTTGATTGGAGAATTTAATTC
Phenotype
MIM: 612786
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CCNY PubMed Links

Gene Details

Gene
CCNY
Gene Name
cyclin Y
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282852.1 Intron NP_001269781.1
NM_001282853.1 Intron NP_001269782.1
NM_001282854.1 Intron NP_001269783.1
NM_145012.5 Intron NP_659449.3
NM_181698.3 Intron NP_859049.2
XM_006717392.3 Intron XP_006717455.1
XM_011519357.2 Intron XP_011517659.1
XM_011519358.1 Intron XP_011517660.1
XM_011519360.2 Intron XP_011517662.1
XM_011519361.1 Intron XP_011517663.1
XM_011519362.1 Intron XP_011517664.1
XM_011519363.1 Intron XP_011517665.1
XM_011519364.1 Intron XP_011517666.1
XM_011519365.2 Intron XP_011517667.1
XM_011519366.1 Intron XP_011517668.1
XM_011519367.1 Intron XP_011517669.1
XM_011519368.2 Intron XP_011517670.1
XM_011519369.2 Intron XP_011517671.1
XM_017015839.1 Intron XP_016871328.1
XM_017015840.1 Intron XP_016871329.1
XM_017015841.1 Intron XP_016871330.1

View Full Product Details