Product Details

SNP ID
rs117645636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:63526959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTAAAATTAGATACTACAGAT[A/G]TTTGATAAGTACTACTTTCCTATGT
Phenotype
MIM: 604503 MIM: 609348
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JMJD1C PubMed Links

Gene Details

Gene
JMJD1C
Gene Name
jumonji domain containing 1C
There are no transcripts associated with this gene.

Gene
REEP3
Gene Name
receptor accessory protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001330.2 Intron NP_001001330.1
XM_011539501.2 Intron XP_011537803.1
XM_017015896.1 Intron XP_016871385.1

View Full Product Details