Product Details

SNP ID
rs138150119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15096419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTGAGAAAAGGAGGCAGATACG[A/T]AAAGAGGAAAAGGAAGGGGGCCCAA
Phenotype
MIM: 603801 MIM: 606116
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACBD7 PubMed Links

Gene Details

Gene
ACBD7
Gene Name
acyl-CoA binding domain containing 7
There are no transcripts associated with this gene.

Gene
C10orf111
Gene Name
chromosome 10 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153244.1 680 Missense Mutation ACG,TCG T136S NP_694976.1
Gene
NMT2
Gene Name
N-myristoyltransferase 2
There are no transcripts associated with this gene.

Gene
RPP38
Gene Name
ribonuclease P/MRP subunit p38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097590.2 680 Intron NP_001091059.1
NM_001265601.1 680 Intron NP_001252530.1
NM_006414.4 680 Intron NP_006405.2
NM_183005.4 680 Intron NP_892117.1
XM_006717363.1 680 Intron XP_006717426.1
XM_006717364.3 680 Intron XP_006717427.1
XM_011519293.1 680 Intron XP_011517595.1
XM_017015479.1 680 Intron XP_016870968.1
XM_017015480.1 680 Intron XP_016870969.1
XM_017015481.1 680 Intron XP_016870970.1

View Full Product Details