Product Details

SNP ID
rs138428691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101064548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGTGACTGGCTGGCTGCAGAT[C/T]CAGGCTGTGCGTCCCAGTGATGAGG
Phenotype
MIM: 609208
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAZALD1 PubMed Links

Gene Details

Gene
KAZALD1
Gene Name
Kazal type serine peptidase inhibitor domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319303.1 1645 Silent Mutation ATC,ATT I102I NP_001306232.1
NM_030929.4 1645 Silent Mutation ATC,ATT I240I NP_112191.2
XM_005270194.3 1645 UTR 3 XP_005270251.1
XM_017016714.1 1645 Silent Mutation ATC,ATT I240I XP_016872203.1
XM_017016715.1 1645 Silent Mutation ATC,ATT I240I XP_016872204.1

View Full Product Details