Product Details

SNP ID
rs138712777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:56358445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCAAATTTACATCTCCAGCAGG[C/T]TGTAGACCAACAGCCTTGGAGAAAT
Phenotype
MIM: 609177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZWINT PubMed Links

Gene Details

Gene
ZWINT
Gene Name
ZW10 interacting kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005413.1 844 Silent Mutation CAA,CAG Q222Q NP_001005413.1
NM_007057.3 844 Silent Mutation CAA,CAG Q269Q NP_008988.2
NM_032997.2 844 Silent Mutation CAA,CAG Q269Q NP_127490.1
XM_017015605.1 844 Missense Mutation AAC,AGC N257S XP_016871094.1

View Full Product Details