Product Details

SNP ID
rs141410181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73910334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGCTCATTATGGAAACTTCCCA[A/G]AAGACCCGTTAACACTTCAATAGGA
Phenotype
MIM: 191840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf55 PubMed Links

Gene Details

Gene
C10orf55
Gene Name
chromosome 10 open reading frame 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001791.2 2148 UTR 3 NP_001001791.2
Gene
PLAU
Gene Name
plasminogen activator, urokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145031.2 2148 Intron NP_001138503.1
NM_001319191.1 2148 Intron NP_001306120.1
NM_002658.4 2148 Intron NP_002649.1
XM_011539866.2 2148 Intron XP_011538168.1

View Full Product Details