Product Details

SNP ID
rs143215831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102919094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCCGCCCTGGGCGCCGGCGGCT[C/T]GGGGTCCACGGGTGGCGCCGTCGGG
Phenotype
MIM: 607803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNNM2 PubMed Links

Gene Details

Gene
CNNM2
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017649.4 802 Missense Mutation TCG,TTG S205L NP_060119.3
NM_199076.2 802 Missense Mutation TCG,TTG S205L NP_951058.1
NM_199077.2 802 Missense Mutation TCG,TTG S205L NP_951059.1
XM_005269933.4 802 Missense Mutation TCG,TTG S205L XP_005269990.1
XM_006717908.2 802 Missense Mutation TCG,TTG S205L XP_006717971.1
XM_011539911.2 802 Missense Mutation TCG,TTG S205L XP_011538213.1
Gene
LOC107984265
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017017027.1 802 Intron XP_016872516.1

View Full Product Details