Product Details

SNP ID
rs144446056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:126425056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTGTAAAAACTAGCTTGTTCT[C/T]AGCTTCCCAAACAGACATATTTATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf90 PubMed Links

Gene Details

Gene
C10orf90
Gene Name
chromosome 10 open reading frame 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004298.2 3074 UTR 3 NP_001004298.2
XM_005269491.1 3074 UTR 3 XP_005269548.1
XM_005269492.2 3074 UTR 3 XP_005269549.1
XM_011539216.1 3074 UTR 3 XP_011537518.1
XM_011539217.2 3074 UTR 3 XP_011537519.1
XM_011539220.2 3074 UTR 3 XP_011537522.1
XM_011539222.2 3074 UTR 3 XP_011537524.1
XM_011539223.2 3074 UTR 3 XP_011537525.1
XM_011539224.2 3074 UTR 3 XP_011537526.1
XM_011539226.1 3074 UTR 3 XP_011537528.1
XM_011539227.2 3074 UTR 3 XP_011537529.1
XM_011539230.2 3074 UTR 3 XP_011537532.1
XM_011539231.1 3074 UTR 3 XP_011537533.1
XM_011539232.1 3074 UTR 3 XP_011537534.1
XM_011539233.1 3074 UTR 3 XP_011537535.1
XM_017015634.1 3074 UTR 3 XP_016871123.1
XM_017015635.1 3074 Intron XP_016871124.1
Gene
LINC00601
Gene Name
long intergenic non-protein coding RNA 601
There are no transcripts associated with this gene.

Gene
LOC728158
Gene Name
hCG2044975
There are no transcripts associated with this gene.

View Full Product Details