Product Details

SNP ID
rs144481604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84176549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTGCCACCGAGTCCCTAGCC[C/G]CAACTCAACAAACCTGAAAGGTAAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C10orf99 PubMed Links

Gene Details

Gene
C10orf99
Gene Name
chromosome 10 open reading frame 99
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207373.2 194 Missense Mutation CCC,CGC P47R NP_997256.1
XM_005269811.3 194 Missense Mutation CCC,CGC P47R XP_005269868.1
Gene
HOST2
Gene Name
human ovarian cancer-specific transcript 2
There are no transcripts associated with this gene.

View Full Product Details