Product Details

SNP ID
rs144811392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87862043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCTGTAAGAATCGGCGGCAGCC[A/T]GCAGGCGGGGAGGCGGGGGCACGTG
Phenotype
MIM: 612105 MIM: 601728
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLLN PubMed Links

Gene Details

Gene
KLLN
Gene Name
killin, p53-regulated DNA replication inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126049.1 1395 Missense Mutation AGG,TGG R149W NP_001119521.1
Gene
PTEN
Gene Name
phosphatase and tensin homolog
There are no transcripts associated with this gene.

View Full Product Details