Product Details

SNP ID
rs147262032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73120117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTCTGGCTCCTCTGCTTCAGAC[A/T]TCAGCACATCAATACCTGGCCCCCC
Phenotype
MIM: 609233
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NUDT13 PubMed Links

Gene Details

Gene
NUDT13
Gene Name
nudix hydrolase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283014.1 457 Silent Mutation ACA,ACT T61T NP_001269943.1
NM_001283015.1 457 Silent Mutation ACA,ACT T61T NP_001269944.1
NM_001283016.1 457 UTR 5 NP_001269945.1
NM_001283017.1 457 UTR 5 NP_001269946.1
NM_001283019.1 457 UTR 5 NP_001269948.1
NM_015901.5 457 Silent Mutation ACA,ACT T61T NP_056985.3
XM_017016033.1 457 Silent Mutation ACA,ACT T61T XP_016871522.1

View Full Product Details