Product Details

SNP ID
rs148109760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73114429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGGTGCTATAGGCTGCTGTCA[A/G]CCTATGTTACTAAGACACGGTGAGT
Phenotype
MIM: 609233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT13 PubMed Links

Gene Details

Gene
NUDT13
Gene Name
nudix hydrolase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283014.1 266 Missense Mutation ACC,GCC T22A NP_001269943.1
NM_001283015.1 266 Missense Mutation ACC,GCC T22A NP_001269944.1
NM_001283016.1 266 UTR 5 NP_001269945.1
NM_001283017.1 266 UTR 5 NP_001269946.1
NM_001283019.1 266 UTR 5 NP_001269948.1
NM_015901.5 266 Missense Mutation ACC,GCC T22A NP_056985.3
XM_017016033.1 266 Missense Mutation ACC,GCC T22A XP_016871522.1

View Full Product Details