Product Details

SNP ID
rs148686867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:52316693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCATAGGAGAAAAGGCTCTCA[C/T]GGACTAGAAATATTCCAGCGTTGTT
Phenotype
MIM: 605189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKK1 PubMed Links

Gene Details

Gene
DKK1
Gene Name
dickkopf WNT signaling pathway inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012242.2 841 Silent Mutation CAC,CAT H229H NP_036374.1
Gene
PRKG1-AS1
Gene Name
PRKG1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details