Product Details

SNP ID
rs148940784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89206975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGAGGGCCGGGCTGCGGGCCCA[A/G]TGCAGCAGCGTCACGGGGAACACAA
Phenotype
MIM: 604551 MIM: 613497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CH25H PubMed Links

Gene Details

Gene
CH25H
Gene Name
cholesterol 25-hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003956.3 340 Silent Mutation CAC,CAT H106H NP_003947.1
Gene
LIPA
Gene Name
lipase A, lysosomal acid type
There are no transcripts associated with this gene.

View Full Product Details