Product Details

SNP ID
rs149097055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70771398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCAGCTCTCTGCCCTCGGCTTC[A/G]ATGTCTTCTTCTCTGTGTTCTGGCT
Phenotype
MIM: 607506 MIM: 612640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS14 PubMed Links

Gene Details

Gene
ADAMTS14
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
There are no transcripts associated with this gene.

Gene
TBATA
Gene Name
thymus, brain and testes associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318241.1 1673 Missense Mutation NP_001305170.1
NM_001318242.1 1673 Missense Mutation NP_001305171.1
NM_001318243.1 1673 Missense Mutation NP_001305172.1
NM_152710.3 1673 Missense Mutation NP_689923.3
XM_017015845.1 1673 UTR 3 XP_016871334.1
XM_017015846.1 1673 UTR 3 XP_016871335.1
XM_017015847.1 1673 UTR 3 XP_016871336.1
XM_017015848.1 1673 Silent Mutation XP_016871337.1
XM_017015849.1 1673 Nonsense Mutation XP_016871338.1
XM_017015850.1 1673 Intron XP_016871339.1
XM_017015851.1 1673 Intron XP_016871340.1
XM_017015852.1 1673 Silent Mutation XP_016871341.1
XM_017015853.1 1673 Missense Mutation XP_016871342.1
XM_017015854.1 1673 Intron XP_016871343.1
XM_017015855.1 1673 Intron XP_016871344.1
XM_017015856.1 1673 UTR 3 XP_016871345.1
XM_017015857.1 1673 Intron XP_016871346.1
XM_017015858.1 1673 UTR 3 XP_016871347.1
XM_017015859.1 1673 Intron XP_016871348.1
XM_017015860.1 1673 Silent Mutation XP_016871349.1
XM_017015861.1 1673 Intron XP_016871350.1
XM_017015862.1 1673 Intron XP_016871351.1

View Full Product Details