Product Details

SNP ID
rs149711267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117197626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGTCTTCTCTGCCATTGAGGA[C/T]GGCCAGGTCCTGGTGGCAGCAGATG
Phenotype
MIM: 613655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK18 PubMed Links

Gene Details

Gene
KCNK18
Gene Name
potassium two pore domain channel subfamily K member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181840.1 138 Silent Mutation GAC,GAT D46D NP_862823.1

View Full Product Details