Product Details

SNP ID
rs150346438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114187378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAGAGGGTTCAGCATATTCAA[A/G]GATTAGATATAGCATAAAGTAGGAG
Phenotype
MIM: 605796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRD1 PubMed Links

Gene Details

Gene
TDRD1
Gene Name
tudor domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198795.1 Intron NP_942090.1
XM_005269978.2 Intron XP_005270035.1
XM_005269979.2 Intron XP_005270036.1
XM_011539959.2 Intron XP_011538261.1
XM_011539960.2 Intron XP_011538262.1
XM_011539961.2 Intron XP_011538263.1
XM_011539962.1 Intron XP_011538264.1
XM_011539963.1 Intron XP_011538265.1
XM_011539964.1 Intron XP_011538266.1
XM_011539966.1 Intron XP_011538268.1
XM_017016414.1 Intron XP_016871903.1
XM_017016415.1 Intron XP_016871904.1

View Full Product Details