Product Details

SNP ID
rs10583719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8921344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAATTCTTGTTCCACTGACAT[AT/-]TGCAAGGCTCTGACTCCGCTTTCTG
Phenotype
MIM: 609191
Polymorphism
AT/-, Insertion/deletion
Allele Nomenclature
Literature Links
AKIP1 PubMed Links

Gene Details

Gene
AKIP1
Gene Name
A-kinase interacting protein 1
There are no transcripts associated with this gene.

Gene
C11orf16
Gene Name
chromosome 11 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020643.2 1960 Silent Mutation ATA,ATT I459I NP_065694.2
XM_017018013.1 1960 Frame Shift Delete ATA,TTG I459L XP_016873502.1

View Full Product Details