Product Details
- SNP ID
-
rs115152574
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:66314529 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCCATTTATTGGAGAAGACCCCA[A/G]CACCCGCCCCCTGAGGTCTTAAGGG
- Phenotype
-
MIM: 606064
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD248
PubMed Links
Gene Details
- Gene
- CD248
- Gene Name
- CD248 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020404.2 |
2516 |
UTR 3 |
|
|
NP_065137.1 |
View Full Product Details