Product Details

SNP ID
rs115927818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59713225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCAATGAGAGGGGGATGCTAAG[A/G]ACGATGAAGCTGATGATATACAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10V1 PubMed Links

Gene Details

Gene
OR10V1
Gene Name
olfactory receptor family 10 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005324.1 621 Silent Mutation GTC,GTT V207V NP_001005324.1

View Full Product Details