Product Details

SNP ID
rs118060724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66975627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGCAGCCGGTGGTGGCTGAGG[C/T]GGTACCAACAGGTATGGCTGTGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf86 PubMed Links

Gene Details

Gene
C11orf86
Gene Name
chromosome 11 open reading frame 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136485.1 345 Missense Mutation CGG,TGG R89W NP_001129957.1

View Full Product Details