Product Details

SNP ID
rs138339957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5968121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGCTTGCAGATGGCCACATAG[C/T]GGTCATAGGCCATGATCATGAATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR56A5 PubMed Links

Gene Details

Gene
OR56A5
Gene Name
olfactory receptor family 56 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146033.1 374 Missense Mutation CAC,CGC H125R NP_001139505.1

View Full Product Details