Product Details

SNP ID
rs140141901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78064097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCATCTGTGTGGATGTGGACCA[C/T]GGGCTGCTGCCGCGGGAGGAGTGGC
Phenotype
MIM: 603845 MIM: 601926
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFC2 PubMed Links

Gene Details

Gene
NDUFC2
Gene Name
NADH:ubiquinone oxidoreductase subunit C2
There are no transcripts associated with this gene.

Gene
NDUFC2-KCTD14
Gene Name
NDUFC2-KCTD14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203260.1 237 Intron NP_001190189.1
NM_001203261.1 237 Intron NP_001190190.1
NM_001203262.1 237 Intron NP_001190191.1
Gene
THRSP
Gene Name
thyroid hormone responsive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003251.3 237 Silent Mutation CAC,CAT H72H NP_003242.1

View Full Product Details