Product Details
- SNP ID
-
rs140938743
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:35139369 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGTGCCGCTGAGCCTGGCGCAGAT[A/C]GGTGAGTGCCCGCCGCAGCCTGGGC
- Phenotype
-
MIM: 107269
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD44
PubMed Links
Gene Details
- Gene
- CD44
- Gene Name
- CD44 molecule (Indian blood group)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000610.3 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_000601.3 |
NM_001001389.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_001001389.1 |
NM_001001390.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_001001390.1 |
NM_001001391.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_001001391.1 |
NM_001001392.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_001001392.1 |
NM_001202555.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_001189484.1 |
NM_001202556.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_001189485.1 |
NM_001202557.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
NP_001189486.1 |
XM_005253231.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253288.1 |
XM_005253232.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253289.1 |
XM_005253233.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253290.1 |
XM_005253234.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253291.1 |
XM_005253235.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253292.1 |
XM_005253238.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253295.1 |
XM_005253239.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253296.1 |
XM_005253240.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_005253297.1 |
XM_006718388.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_006718451.1 |
XM_006718390.3 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_006718453.1 |
XM_011520482.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518784.1 |
XM_011520483.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518785.1 |
XM_011520484.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518786.1 |
XM_011520485.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518787.1 |
XM_011520486.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518788.1 |
XM_011520487.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518789.1 |
XM_011520488.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518790.1 |
XM_011520489.2 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_011518791.1 |
XM_017018583.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_016874072.1 |
XM_017018584.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_016874073.1 |
XM_017018585.1 |
227 |
Silent Mutation |
ATA,ATC |
I22I |
XP_016874074.1 |
- Gene
- LOC100507144
- Gene Name
- uncharacterized LOC100507144
There are no transcripts associated with this gene.
View Full Product Details