Product Details

SNP ID
rs141261155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9095899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTTGTGAAGAAGGTGGAAACGC[G/A]AGATGGCAAGCTAGTGTCCAAGTCC
Phenotype
MIM: 611747
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
KRT8P41 PubMed Links

Gene Details

Gene
KRT8P41
Gene Name
keratin 8 pseudogene 41
There are no transcripts associated with this gene.

Gene
MIR5691
Gene Name
microRNA 5691
There are no transcripts associated with this gene.

Gene
SCUBE2
Gene Name
signal peptide, CUB domain and EGF like domain containing 2
There are no transcripts associated with this gene.

View Full Product Details