Product Details

SNP ID
rs143043362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56664050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGCAGCCATGGCCTATGGTC[A/G]TTTTGTGGCCATTTGTCGACCCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5AR1 PubMed Links

Gene Details

Gene
OR5AR1
Gene Name
olfactory receptor family 5 subfamily AR member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004730.1 365 Missense Mutation CAT,CGT H122R NP_001004730.1

View Full Product Details