Product Details

SNP ID
rs145434076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56417335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTATCCAAGGCTTTCTTTGAGG[C/T]ATCTTTCACTTCTTTGTTCCTTAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5R1 PubMed Links

Gene Details

Gene
OR5R1
Gene Name
olfactory receptor family 5 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004744.1 898 Missense Mutation ACC,GCC T300A NP_001004744.1

View Full Product Details