Product Details
- SNP ID
-
rs146027803
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:104887276 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAATTTGGGAAATGAAGACAGAGC[C/T]ATTTGTTTCATGTCTCCAAGAAACA
- Phenotype
-
MIM: 608633
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CASP12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142269030] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASP12
- Gene Name
- caspase 12 (gene/pseudogene)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001191016.2 |
1103 |
Missense Mutation |
AGC,GGC |
S283G |
NP_001177945.2 |
View Full Product Details